Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MMKLKSNQTR TYDGDGYKKR AACLCFRSES EEEVLLVSSS RHPDRWIVPG GGMEPEEEPS VAAVREVCEE AGVKGTLGRL VGIFENQERK HRTYVYVLIV TEVLEDWEDS VNIGRKREWF KIEDAIKVLQ YHKPVQASYF ETLRQGYSAN NGTPVVATTY SVSAQSSMSG IR
NUDT3 belongs to the MutT, or Nudix, protein family. Nudix proteins act as homeostatic checkpoints at important stages in nucleoside phosphate metabolic pathways, guarding against elevated levels of potentially dangerous intermediates, like 8-oxo-dGTP, which promotes AT-to-CG transversions (Safrany et al., 1998 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ap6A hydrolase; diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 1; Diadenosine hexaphosphate hydrolase; Diphosphoinositol polyphosphate phosphohydrolase 1; DIPP-1; Endopolyphosphatase; m7GpppN-mRNA hydrolase; m7GpppX diphosphatase; Nucleoside diphosphate-linked moiety X motif 3; nudix (nucleoside diphosphate linked moiety X)-type motif 3; Nudix motif 3
Gene Aliases: DIPP; DIPP-1; DIPP1; NUDT3
UniProt ID: (Human) O95989
Entrez Gene ID: (Human) 11165
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support