Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEEKPGQPQ PQHHHSHHHP HHHPQQQQQQ PHHHHHYYFY NHSHNHHHHH HHQQPHQYLQ HGAEGSPKAQ PKPLKHEQKH TLQQHQETPK KKTGYGELNG NAGEREISLK NLSSDEATNP ISRVLNGNQQ VVDTSLKQTV KANTFGKAGI KTKNFIQKNS MDKKNGKSYE NKSGENQSVD KSDTIPIPNG VVTNNSGYIT NGYMGKGADN DGSGSESGYT TPKKRKARRN SAKGCENLNI VQDKIMQQET SVPTLKQGLE TFKPDYSEQK GNRVDGSKPI WKYETGPGGT SRGKPAVGDM LRKSSDSKPG VSSKKFDDRP KGKHASAVAS KEDSWTLFKP PPVFPVDNSS A (1-350 aa encoded by BC108307)
NUFIP2 binds RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 82 kDa FMRP-interacting protein; 82-FIP; 82-kD FMRP Interacting Protein; Cell proliferation-inducing gene 1 protein; FMR1-interacting protein NUFIP2; FMRP-interacting protein 2; Fragile X Mental Retardation Protein; Nuclear FMR1-interacting protein 2; nuclear fragile X mental retardation protein interacting protein 2; proliferation-inducing gene 1
Gene Aliases: 182-FIP; 82-FIP; FIP-82; KIAA1321; NUFIP2; PIG1
UniProt ID: (Human) Q7Z417
Entrez Gene ID: (Human) 57532
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support