Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
NUFIP2 binds RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 82 kDa FMRP-interacting protein; 82-FIP; 82-kD FMRP Interacting Protein; Cell proliferation-inducing gene 1 protein; FMR1-interacting protein NUFIP2; FMRP-interacting protein 2; Fragile X Mental Retardation Protein; Nuclear FMR1-interacting protein 2; nuclear fragile X mental retardation protein interacting protein 2; nuclear fragile X mental retardation-interacting protein 2; proliferation-inducing gene 1
Gene Aliases: 1110001M19Rik; 182-FIP; 82-FIP; 9530056D24Rik; AI256692; AI461731; FIP-82; KIAA1321; mKIAA1321; NUFIP2; PIG1
UniProt ID: (Human) Q7Z417, (Mouse) Q5F2E7
Entrez Gene ID: (Human) 57532, (Mouse) 68564, (Rat) 681371
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support