Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAATSKEVAR LETLVRKAGE QQETASRELV KEPARAGDRQ PEWLEEQQGR QFCSTQAALQ AMEREAEQMG NELERLRAAL MESQGQQQEE RGQQEREVAR LTQERGRAQA DLALEKAARA ELEMRLQNAL NEQRVEFATL QEALAHALTE KEGKDQELAK LRGLEAAQIK ELEELRQTVK QLKEQLAKKE KEHASGSGAQ SEAAGRTEPT GPKLEALRAE VSKLEQQCQK QQEQADSLER SLEAERASRA ERDSALETLQ GQLEEKAQEL GHSQSALASA QRELAAFRTK VQDHSKAEDE WKAQVARGRQ EAERKNSLIS SLEEEVSILN RQVLEKEGES KELKRLVMAE SEKSQKLEER LRLLQAETAS NSARAAERSS ALREEVQSLR EEAEKQRVAS ENLRQELTSQ AERAEELGQE LKAWQEKFFQ KEQALSTLQL EHTSTQALVS ELLPAKHLCQ QLQAEQAAAE KRHREELEQS KQAAGALRAE LLRAQRELGE LIPLRQKVAE QERTAQQLRA EKASYAEQLS MLKKAHGLLA EENRGLGERA NLGRQFLEVE LDQAREKYVQ ELAAVRADAE TRLAEVQREA QSTARELEVM TAKYEGAKVK VLEERQRFQE ERQKLTAQVE ELSKKLADSD QASKVQQQKL KAVQAQGGES QQEAQRLQAQ LNELQAQLSQ KEQAAEHYKL QMEKAKTHYD AKKQQNQELQ EQLRSLEQLQ KENKELRAEA ERLGHELQQA GLKTKEAEQT CRHLTAQVRS LEAQVAHADQ QLRDLGKFQV ATDALKSREP QAKPQLDLSI DSLDLSCEEG TPLSITSKLP RTQPDGTSVP GEPASPISQR LPPKVESLES LYFTPIPARS QAPLESSLDS LGDVFLDSGR KTRSARRRTT QIINITMTKK LDVEEPDSAN SSFYSTRSAP ASQASLRATS STQSLARLGS PDYGNSALLS LPGYRPTTRS SARRSQAGVS SGAPPGRNSF YMGTCQDEPE QLDDWNRIAE LQQRNRVCPP HLKTCYPLES RPSLSLGTIT DEEMKTGDPQ ETLRRASMQP IQIAEGTGIT TRQQRKRVSL EPHQGPGTPE SKKATSCFPR PMTPRDRHEG RKQSTTEAQK KAAPASTKQA DRRQSMAFSI LNTPKKLGNS LLRRGASKKA LSKASPNTRS GTRRSPRIAT TTASAATAAA IGATPRAKGK AKH
NuMA (nuclear mitotic apparatus) is a long coil-coiled protein that plays a role in the interphase and mitosis phases of a cell cycle. In interphase, it has been hypothesized to play a structural role in the nucleoskeleton that may be important in nuclear organization and functions. During mitosis, it is associated with spindle microtubule organization and chromosome positioning.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centrophilin stabilizes mitotic spindle in mitotic cells; NMP-22; Nuclear matrix protein-22; Nuclear mitotic apparatus protein; Nuclear mitotic apparatus protein 1; NuMA protein; SP-H antigen; structural nuclear protein
Gene Aliases: 6720401E04Rik; AA764025; AL022610; AU014979; NMP-22; NMP22; NUMA; NUMA1
UniProt ID: (Human) Q14980, (Mouse) E9Q7G0
Entrez Gene ID: (Human) 4926, (Mouse) 101706
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support