Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive control: Hela cell lysate, 293T cell lysate, HepG2 cell lysate, Jurkat cell lysate, PC-3M cell lysate, A549 cell lysate, MCF-7 cell lysate, PC-12 cell lysate, RAW264.7 cell lysate, NIH/3T3 cell lysate, mouse testis tissue lysate.
Predicted band size: 106 kDa
Subcellular Location: Nucleus membrane, Nucleus, nuclear pore, Chromosome, centromere, kinetochore.
The nuclear pore complex (NPC) is a protein assembly localized at the nuclear rim and mediates macromolecular transport between the nucleus and the cytoplasm. The mammalian nucleoporin (NUP)-107 is part of the hetero-oligomeric complex that also contains NUP160, NUP133, NUP96, and mammalian homolog of yeast sec13p. While the majority of the NUP107-160 nuclear pore sub-complex localizes to the nuclear pore, a small fraction is observed at kinetochores and pro-metaphase spindle poles in mitotic cells in association with proteins such as Mad1, Mad2, Bub3 and Cdc20. Immunodepletion of the NUP107-160 complex resulted in defective spindle assembly indicating that it has multiple functions. NUP107 has recently been identified as an HIV dependency factor (HDF), suggesting that NUP107 may be an important drug target in HIV treatment. Multiple isoforms of NUP107 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 107 kDa nucleoporin; nuclear pore complex; Nuclear pore complex protein Nup107; nucleoporin 107kDa; Nucleoporin Nup107; p105
Gene Aliases: AW541137; C76801; NPHS11; NUP107; NUP84
UniProt ID: (Human) P57740, (Mouse) Q8BH74
Entrez Gene ID: (Human) 57122, (Rat) 116555, (Mouse) 103468
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support