Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAFAVEPQG PALGSEPMML GSPTSPKPGV NAQFLPGFLM GDLPAPVTPQ PRSISGPSVG VMEMRSPLLA GGSPPQPVVP AHKDKSGAPP VRSIYDDISS PGLGSTPLTS RRQPNISVMQ SPLVGVTSTP GTGQSMFSPA SIGQPRKTTL SPAQLDPFYT QGDSLTSEDH LDDSWVTVFG FPQASASYIL LQFAQYGNIL KHVMSNTGNW MHIRYQSKLQ ARKALSKDGR IFGESIMIGV KPCIDKSVME SSDRCALSSP SLAFTPPIKT LGTPTQPGST PRISTMRPLA TAYKASTSDY QVISDRQTPK KDESLVSKAM EYMFGW
Nucleoporin Nup35 or Mitotic phosphoprotein 44 is a nuclear protein known to be present on nuclear pore complex and is tightly associated with the nuclear membrane and lamina. This RNA associated protein belongs to the Nup53 family and has a MPPN (mitotic phosphoprotein N' end) domain. It has a small number of scattered phenylalanine-glycine repeats, but not the large domains seen in other nucleoporins. Though the exact function of NUP35 is yet to be revealed, it may function as a component of the nuclear pore complex (NPC) which are collectively referred to as nucleoporins (NUPs). It also may play the role of both NPC structural components and of docking or interaction partners for transiently associated nuclear transport factors, and may also play an important part in the association of MAD1, a transcriptional repressor with the NPC. NUP35 is widely expressed in most of the tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 35 kDa nucleoporin; Mitotic phosphoprotein 44; MP-44; Nuclear pore complex protein Nup53; nucleoporin 35kDa; Nucleoporin NUP35; Nucleoporin NUP53
Gene Aliases: MP-44; MP44; NP44; NUP35; NUP53
UniProt ID: (Human) Q8NFH5
Entrez Gene ID: (Human) 129401
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support