Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAFNFGAPS GTSGTAAATA APAGGFGGFG TTSTTAGSAF SFSAPTNTGT TGLFGGTQNK GFGFGTGFGT TTGTSTGLGT GLGTGLGFGG FNTQQQQQTT LGGLFSQPTQ APTQSNQLIN TASALSAPTL LGDERDAILA KWNQLQAFWG TGKGYFNNNI PPVEFTQENP FCRFKAVGYS CMPSNKDEDG LVVLVFNKKE TEIRSQQQQL VESLHKVLGG NQTLTVNVEG TKTLPDDQTE VVIYVVERSP NGTSRRVPAT TLYAHFEQAN IKTQLQQLGV TLSMTRTELS PAQIKQLLQN PPAGVDPIIW EQAKVDNPDS EKLIPVPMVG FKELLRRLKV QDQMTKQHQT RL (1-351 aa encoded by BC012559)
The nuclear envelope creates distinct nuclear and cytoplasmic compartments in eukaryotic cells. It consists of two concentric membranes perforated by nuclear pores, large protein complexes that form aqueous channels to regulate the flow of macromolecules between the nucleus and the cytoplasm. These complexes are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. This gene encodes a member of the phe-gly repeat-containing nucleoporin subset.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 54 kDa nucleoporin; Nuclear pore complex protein Nup54; nucleoporin 54kD; nucleoporin 54kDa; Nucleoporin Nup54; Nucleoporin p54
Gene Aliases: 3110079L04Rik; 54kDa; NUP54
UniProt ID: (Human) Q7Z3B4, (Rat) P70582, (Mouse) Q8BTS4
Entrez Gene ID: (Human) 53371, (Rat) 53372, (Mouse) 269113
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support