Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SGIRSSRLEE DDGDVAMSDA QDGPRVRYNP YTTRPNRRGD TWHDRDRIHV TVRRDRAPPE RGGAGTSQDG TSKNWFKITI PYGRKYDKAW LLSMIQSKCS VPFTPIEFHY ENTRAQFFVE DASTASALKA VNYKILDREN RRISIIINSS APPHTILNEL KPEQVEQLKL IMSKRYDGSQ QALDLKGLRS DPDLVAQNID VVLNRRSCMA ATLRIIEENI PELLSLNLSN NRLYRLDDMS SIVQKAPNLK ILNLSGNELK SERELDKIKG LKLEELWLDG NSLCDTFRDQ STYISAIRER FPKLL
The constitutive transport element (CTE) of type D retroviruses serves as a signal of nuclear export for unspliced viral RNAs. TAP (also known as NXF1) mediates the export of CTE-containing simian type D retroviral RNAs through binding directly to the CTE. TAP is associated with a recognized mRNA export pathway and is a member of the multigene family of NXF proteins. NXF proteins belong to an evolutionarily conserved family of proteins, which are characterized by a leucine-rich-repeat domain (LRR) followed by a region known as the Nuclear Transport Factor 2 (NTF2)-like domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mRNA export factor TAP; Nuclear RNA export factor 1; tip associating protein; Tip-associated protein; Tip-associating protein; Vdp
Gene Aliases: MEX67; NXF1; TAP
UniProt ID: (Human) Q9UBU9
Entrez Gene ID: (Human) 10482
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support