Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MCSTLKKCGT YRTEVAECHD HGSTFQGRKK GGSSFRDNFD KRSCHYEHGG YERPPSHCQE NDGSVEMRDV HKDQQLRHTP YSIRCERRMK WHSEDEIRIT TWRNRKPPER KMSQNTQDGY TRNWFKVTIP YGIKYDKAWL MNSIQSHCSD RFTPVDFHYV RNRACFFVQD ASAASALKDV SYKIYDDENQ KICIFVNHST APYSVKNKLK PGQMEMLKLT MNKRYNVSQQ ALDLQNLRFD PDLMGRDIDI ILNRRNCMAA TLKIIERNFP ELLSLNLCNN KLYQLDGLSD ITEKAPKVKT LNLSKNKLES AWELGKVKGL KLEELWLEGN PLCSTFSDQS AYVSAIRDCF PKLLRLDGRE LSAPVIVDID SSETMKPCKE NFTGSETLKH LVLQFLQQYY SIYDSGDRQG LLGAYHDEAC FSLAIPFDPK DSAPSSLCKY FEDSRNMKTL KDPYLKGELL RRTKRDIVDS LSALPKTQHD LSSILVDVWC QTERMLCFSV NGVFKEVEGQ SQGSVLAFTR TFIATPGSSS SLCIVNDELF VRDASPQETQ SAFSIPVSTL SSSSEPSLSQ EQQEMVQAFS AQSGMKLEWS QKCLQDNEWN YTRAGQAFTM LQTEGKIPAE AFKQIS
This gene is one of a family of nuclear RNA export factor genes. It encodes a protein that is involved in mRNA export, is located in the nucleoplasm, and is associated with the nuclear envelope. Alternative splicing seems to be a common mechanism in this gene family. Two variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 39; CT39; Nuclear RNA export factor 2; t-complex protein 11 homolog; TAP-like protein 2; TAPL-2
Gene Aliases: CT39; NXF2; NXF2B; TAPL-2; TAPL2; TCP11X2
UniProt ID: (Human) Q9GZY0
Entrez Gene ID: (Human) 56001
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support