Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ADVSLHRNP ATLKLWNKYR ISNIPSLIFL DATTGKVVCR NGLLVIRDDP EGLEFPWGPK PFREVIAGPL LRNNGQSLES SSLEGSHVGV YFSAHWCPPC RSLTRVLVES YRKIKEAGQN FEIIFVSADR SEESFKQYFS EMPWLAVPYT DEARRSRLNR LYGIQGIPTL IMLDPQGEVI TRQGRVEVLN DEDCREFPWH PKPVLELSDS NAAQLNEGPC LVLFVDSEDD GESEAAKQLI QPIAEKIIAK YKAKEEEAPL LFFVAGEDDM TDSLRDYTNL PEAAPLLTIL DMSARAKYVM DVEEITPAIV EAFVNDFLAE KLKPEPI (110-435 aa encoded by BC009327 )
This gene encodes a member of the thioredoxin superfamily, a group of small, multifunctional redox-active proteins. Members of this family are characterized by a conserved active motif called the thioredoxin fold that catalyzes disulfide bond formation and isomerization. The encoded protein acts a redox-dependent regulator of the Wnt signaling pathway and is involved in cell growth and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nucleoredoxin; nucleoredoxin 1; Protein Red-1
Gene Aliases: Gn25; l11Jus13; NRX; NXN; TRG-4
UniProt ID: (Human) Q6DKJ4, (Mouse) Q5H8U0
Entrez Gene ID: (Human) 64359, (Mouse) 18230, (Rat) 360577
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support