Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRLLPEWFLL LFGPWLLRKA VSAQIPESGR PQYLGLRPAA AGAGAPGQQL PEPRSSDGLG VGRAWSWAWP TNHTGALARA GAAGALPAQR TKRKPSIKAA RAKKIFGWGD FYFRVHTLKF SLLVTGKIVD HVNGTFSVYF RHNSSSLGNL SVSIVPPSKR VEFGGVWLPG PVPHPLQSTL ALEGVLPGLG PPLGMAAAAA GPGLGGSLGG ALAGPLGGAL GVPGAKESRA FNCHVEYEKT NRARKHRPCL YDPSQVCFTE HTQSQAAWLC AKPFKVICIF VSFLSFDYKL VQKVCPDYNF QSEHPYFG
Members of the Neurexophilin family are neuropeptide-like glycoproteins that undergo proteolytic processing after synthesis. As secreted proteins, Neurexophilins are thought to function as signaling molecules which specifically bind to target proteins, such as neurexin I-alpha (a protein that promotes adhesion between dendrites and axons), and are essential for proper neurotransmitter release. Neurexophilins contain five common domains including a variable N-terminal domain, a highly conserved N-glycosylated central domain, a short linker region and a cysteine-rich conserved C-terminal domain. Neurexophilin-4, also known as NXPH4 or NPH4, is a 308 amino acid secreted protein belonging to the neurexophilin family. Neurexophilin-4 is expressed in brain, spleen and testis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alternative protein NXPH4; Neurexophilin-4
Gene Aliases: NPH4; NXPH4; UNQ1928/PRO4403
UniProt ID: (Human) O95158
Entrez Gene ID: (Human) 11247
Molecular Function:
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support