Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SLFGQKEVR KDPSQESEEN TAAVRRNGLH TVPTEMQFLA STEAKDHPGE TEVPLDLPAD SRKEDEYRRN (161-229 aa encoded by BC035966)
Mammalian sodium-dicarboxylate cotransporters transport succinate and other Krebs cycle intermediates. They fall into 2 categories based on their substrate affinity: low affinity and high affinity. Both the low- and high-affinity transporters play an important role in the handling of citrate by the kidneys. The protein encoded by this gene represents the high-affinity form. Alternatively spliced transcript variants encoding different isoforms have been found for this gene, although the full-length nature of some of them have not been characterized yet.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hNaDC3; mNaDC3; Na(+)-coupled carboxylate transporter 3; Na(+)/dicarboxylate cotransporter 3; NaC3; NaDC-3; SLC13A3; sodium-dependent high affinity dicarboxylate transporter 3; Sodium-dependent high-affinity dicarboxylate transporter 2; Solute carrier family 13 member 3
Gene Aliases: NaDC-3; NADC3; SDCT2; SLC13A3
UniProt ID: (Human) Q8WWT9, (Mouse) Q91Y63
Entrez Gene ID: (Human) 64849, (Mouse) 114644
Molecular Function:
transporter
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support