Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to NEO1.
The protein encoded by this gene mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin-associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains - a RING, a B-box type 1 and a B-box type 2 - and a coiled-coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HsT17534; Immunoglobulin superfamily DCC subclass member 2; NEO; Neogenin; neogenin homolog 1; NGNDKFZp547A066
Gene Aliases: 2610028H22Rik; AI327052; D930014N22Rik; IGDCC2; NEO1; NGN; NTN1R2
UniProt ID: (Human) Q92859, (Mouse) P97798
Entrez Gene ID: (Human) 4756, (Rat) 81735, (Mouse) 18007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support