Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
This antibody detects human, mouse, rat Neurofascin in Western blots and rat Neurofascin in direct ELISAs.
Reconstitute at 0.2 mg/mL in sterile PBS.
Neurofascin encodes an L1 family immunoglobulin cell adhesion molecule with multiple IGcam and fibronectin domains. The protein functions in neurite outgrowth, neurite fasciculation, and organization of the axon initial segment (AIS) and nodes of Ranvier on axons during early development. Both the AIS and nodes of Ranvier contain high densities of voltage-gated Na+ (Nav) channels which are clustered by interactions with cytoskeletal and scaffolding proteins including this protein, gliomedin, ankyrin 3 (ankyrin-G), and betaIV spectrin. This protein links the AIS extracellular matrix to the intracellular cytoskeleton. Neurofascin undergoes extensive alternative splicing, and the full-length nature of some variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686P2250; KIAA0756FLJ46866; Neurofascin; neurofascin homolog
Gene Aliases: AA387016; D430023G06Rik; KIAA0756; mKIAA0756; NF; NFASC; NRCAML
UniProt ID: (Human) Q68DH3, (Rat) P97685, (Mouse) Q810U3
Entrez Gene ID: (Human) 23114, (Rat) 116690, (Mouse) 269116
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support