Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
FIGURE: 1 / 2
Reconstitution: 1 X 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C, protected from the light, for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 X g 5 min).
Tachykinin Receptor 1, a member of the Tachykinin Receptor subfamily, is involved in the mediation of phosphatidylinositol metabolism by the tachykinin substance P, also referred to as neurokinin 1. It is associated with the management of depression, anxiety, and stress. It is also involved in regulating behavioral responses, neuronal survival, and cardiovascular and respiratory functions. The tachykinin receptor 1 is implicated in the progression of rheumatoid arthritis and osteoarthritis. Two variants of the tachykinin receptor 1 are produced by alternative splicing. Tachykinin receptor 1 has been reported primarily in brain, and in blood lymphocytes, nose, small intestine, and stomach. ESTs have been isolated from brain and spleen libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neurokinin 1 receptor; neurokinin receptor 1; NK-1 receptor; NK-1R; Nk1 receptor; SPR; substance P receptor; Substance-P receptor; Tachykinin receptor 1; tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor)
Gene Aliases: NK1R; NKIR; SPR; TAC1R; TACR1
UniProt ID: (Human) P25103, (Mouse) P30548, (Rat) P14600
Entrez Gene ID: (Human) 6869, (Mouse) 21336, (Rat) 24807
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support