Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Mouse: 99% identity (125/126 amino acids identical). Human: 99% identity (125/126 amino acids identical) >40% identity with Neuroligin-2 and -3.
2 µg/mL of MA5-45687 was sufficient for detection of Neuroligin-1 in 20 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 120kDa. Does not cross-react with other Neuroligins.
This antibody was formerly sold as clone S97A-31.
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC45115; Neuroligin I; Neuroligin-1; NLG 1
Gene Aliases: 6330415N05Rik; BB179718; KIAA1070; mKIAA1070; NL1; Nlg1; NLGN1
UniProt ID: (Human) Q8N2Q7, (Mouse) Q99K10, (Rat) Q62765
Entrez Gene ID: (Human) 22871, (Mouse) 192167, (Rat) 116647
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support