Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total NLGN2.
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuroligin-2; ortholog of human and rat neuroligin 2 NLGN2
Gene Aliases: KIAA1366; NL2; NLGN2
UniProt ID: (Human) Q8NFZ4, (Rat) Q62888, (Mouse) Q69ZK9
Entrez Gene ID: (Human) 57555, (Rat) 117096, (Mouse) 216856
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support