Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Homophilic cell adhesion molecule that promotes axonal growth. May play a role in nerve regeneration and in the formation and function of other tissues. Cell adhesion requires divalent cations.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hNINJ1; Nerve injury-induced protein 1; nerve injury-induced protein-1; Ninjurin-1
Gene Aliases: AU024536; NIN1; NINJ1; NINJURIN
UniProt ID: (Human) Q92982, (Mouse) O70131, (Rat) P70617
Entrez Gene ID: (Human) 4814, (Mouse) 18081, (Rat) 25338
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support