Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
FIGURE: 1 / 1
Reconstitution: 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. The nociceptin receptor is a G-protein coupled opioid receptor that functions as receptor for the endogenous neuropeptide nociceptin. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G proteins) and modulates the activity of down-stream effectors. Signaling via G proteins mediates inhibition of adenylate cyclase activity and calcium channel activity. Arrestins modulate signaling via G proteins and mediate the activation of alternative signaling pathways that lead to the activation of MAP kinases. Plays a role in modulating nociception and the perception of pain. Plays a role in the regulation of locomotor activity by the neuropeptide nociceptin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: K3 opiate receptor; Kappa-type 3 opioid receptor; kappa3-related opioid receptor; KOR-3; LC132 Receptor-Like; N/OFQ receptor; Nociceptin receptor; nociceptin receptor ORL1; nociceptin/ orphaninFQ receptor; nociceptin/orphanin FQ receptor; opiate receptor-like 1; opioid receptor-like 1; OPPRL1; ORGC; Orphanin FQ receptor; orphanin FQ receptor-a; orphanin FQ receptor-b; orphanin FQ receptor-e; peptide receptor; ROR-C; seven transmembrane G protein-coupled receptor; XOR1
Gene Aliases: KOR-3; KOR3; LC132; MOR-C; morc; NOCIR; NOP; NOPr; OFQR; OOR; Oprl; OPRL1; ORGC; ORL1; XOR1
UniProt ID: (Human) P41146, (Mouse) P35377, (Rat) P35370
Entrez Gene ID: (Human) 4987, (Mouse) 18389, (Rat) 29256
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support