Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Additional Information: This antibody is expected to recognize reported isoform A (NP_065393.1) and isoform E (NP_997404.1).
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
NogoA is a member of a family of integral membrane proteins termed reticulons that are thought to be involved in numerous disorders including neurodegenerative diseases. Reticulon proteins are known to regulate many cellular processes and interact with multiple proteins and receptors such as BACE. NogoA was initially identified as a myelin-associated neurite outgrowth inhibitor. It is highly expressed in oligodendrocytes in the white matter of the CNS, blocking its activity with antibodies or other factors results in improved axon regrowth and functional recovery in experimental CNS lesion models. NogoA has also been suggested to play a role in neurodegenerative diseases such as Amyotrophic lateral sclerosis, in which case NogoA is found at elevated levels in postmortem muscular samples, and multiple sclerosis (MS), in which case autoantibodies to NogoA have been found in serum and cerebrospinal fluid in MS patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Foocen; Human NogoA; My043 protein; neurite growth inhibitor 220; Neurite outgrowth inhibitor; Neuroendocrine-specific protein; Neuroendocrine-specific protein C homolog; Nogo protein; NSP; reticulon 5; Reticulon-4; Reticulon-5; RTN-x
Gene Aliases: ASY; KIAA0886; My043; Nbla00271; Nbla10545; NI220/250; NOGO; NOGO-A; Nogo-B; Nogo-C; NOGOC; NSP; NSP-CL; RTN-X; RTN4; RTN4-A; RTN4-B1; RTN4-B2; RTN4-C; SP1507
UniProt ID: (Human) Q9NQC3
Entrez Gene ID: (Human) 57142
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support