Invitrogen
The MA1-7405 antibody reacts with the capsid protein from Norovirus type G1.
MA1-7405 has been successfully used in ELISA applications.
The MA1-7405 antibody was raised against the Norovirus Viral capsid.
Protein Aliases: Norwalk virus