Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VESLESLYF TPIPARSQAP LESSLDSLGD VFLDSGRKTR SARRRTTQII NITMTKKLDV EEPDSANSSF YSTRSAPASQ ASLRATSSTQ SLARLGSPDY GNSALLSLPG YRPTTRSSAR RSQAGVSSGA PPGRNSFYMG TCQDEPEQLD DWNRIAELQQ RNRVCPPHLK TCYPLESRPS LSLGTITDEE MKTGDPQETL RRASMQPIQI AEGTGITTRQ QRKRVSLEPH QGPGTPESKK ATSCFPRPMT PRDRHEGRKQ STTEAQKKAA PASTKQADRR QSMAFSILNT PKKLGNSLLR RGASKKALSK ASPNTRSGTR RSPRIATTTA SAATAAAIGA TPRAKGKAKH (845-1193 aa encoded by BC013023)
NuMA (nuclear mitotic apparatus) is a long coil-coiled protein that plays a role in the interphase and mitosis phases of a cell cycle. In interphase, it has been hypothesized to play a structural role in the nucleoskeleton that may be important in nuclear organization and functions. During mitosis, it is associated with spindle microtubule organization and chromosome positioning.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centrophilin stabilizes mitotic spindle in mitotic cells; NMP-22; Nuclear matrix protein-22; Nuclear mitotic apparatus protein; Nuclear mitotic apparatus protein 1; NuMA protein; SP-H antigen; structural nuclear protein
Gene Aliases: 6720401E04Rik; AA764025; AL022610; AU014979; NMP-22; NMP22; NUMA; NUMA1
UniProt ID: (Human) Q14980, (Mouse) E9Q7G0
Entrez Gene ID: (Human) 4926, (Rat) 308870, (Mouse) 101706
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support