Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-5' exonuclease NME7; cilia and flagella associated protein 67; NDK 7; NDP kinase 7; nm23-H7; non-metastatic cells 7, protein expressed in (nucleoside-diphosphate kinase); nucleoside diphosphate kinase 7; Nucleoside diphosphate kinase homolog 7; nucleoside-diphosphate kinase; nucleoside-diphosphate kinase 7; Protein kinase NME7
Gene Aliases: CFAP67; D530024H21Rik; MN23H7; NDK 7; NDK7; nm23-H7; Nm23-M7; Nm23-r7; NME7
UniProt ID: (Human) Q9Y5B8, (Mouse) Q9QXL8, (Rat) Q9QXL7
Entrez Gene ID: (Human) 29922, (Mouse) 171567, (Rat) 171566
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support