Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 5
Description: This 12.14 monoclonal antibody reacts with mouse Nur77 (also known as NR4A1, TR3, NGFI-B, or NAK1), an inducible orphan nuclear receptor. Expressed in thymocytes and T cell lines, Nur77 promotes apoptosis and plays a role in thymocyte negative selection. Additionally, Nur77 has been shown to be critical for steroid biosynthesis in Leydig cells as well as for the effects of dopamine. In addition, Nur77 has been shown to interact with FoxP3 in regulatory T cells. However, our results with this antibody do not correlate with this observation.
Applications Reported: This 12.14 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.
Applications Tested: This 12.14 antibody has been tested by intracellular staining followed by flow cytometric analysis of stimulated mouse thymocytes using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523-00) and protocol. Please refer to "Staining Intracellular Antigens for Flow Cytometry, Protocol B: One step protocol for intracellular (nuclear) proteins" located at www.thermofisher.com/flowprotocols . This may be used at less than or equal to 1.0 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
eFluor 450 is an alternative to Pacific Blue. eFluor 450 emits at 446 nm and is excited with the violet laser line (405 nm). Please make sure that your instrument is capable of detecting this fluorochrome.
Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.
NR4A1 is a member of the steroid-thyroid hormone-retinoid receptor superfamily. NGFI-B-alpha is expressed in brain, adrenal, thyroid, liver, testis, ovary, thymus, muscle, lung and prostate. Its expression is induced in response to various stress stimuli and growth factors. The encoded protein acts as a nuclear transcription factor involved in mediating apoptotic signaling in thymocytes and tumor cells, as well as having a role in signaling in the hypothalamic-pituitary axis. Translocation of the protein from the nucleus to mitochondria induces apoptosis. Three transcript variants encoding two distinct isoforms have been identified for this NR4A1. Variant (3), also known as TRC beta, contains multiple differences compared to variant 1. The resulting isoform b contains a shorter and distinct C-terminus compared to isoform a. RXR has been shown to be a partner for NR4A1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hormone receptor; MGC9485; Nuclear hormone receptor NUR/77; Nuclear protein N10; Nuclear receptor subfamily 4 group A member 1; Orphan nuclear receptor HMR; Orphan receptor tr3
Gene Aliases: Gfrp; GFRP1; Hbr-1; Hbr1; Hmr; N10; NGFI-B; NGFIB; NP10; Nr4a1; Nur77; TIS1; TR3
UniProt ID: (Mouse) P12813
Entrez Gene ID: (Mouse) 15370
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support