Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: mouse brain, rat brain
Predicted Reactivity: Human (98%), Pig (98%), Rhesus Monkey (98%), Bovine (98%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The Nuclear receptor related 1 protein (Nurr1) also known as NR4A2 (nuclear receptor subfamily 4, group A, member 2) is a protein that in humans is encoded by the NR4A2 gene. NURR1 is a member of the nuclear receptor family of intracellular transcription factors.Nurr1 plays a key role in the maintenance of the dopaminergic system of the brain. Mutations in this gene have been associated with disorders related to dopaminergic dysfunction, including Parkinson's disease, schizophrenia, and manic depression. Misregulation of this gene may be associated with rheumatoid arthritis. Four transcript variants encoding four distinct isoforms have been identified for this gene. Additional alternate splice variants may exist, but their full-length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HZF3; NGFI-B beta; Nuclear orphan receptor HZF-3; Nuclear receptor subfamily 4 group A member 2; nuclear receptor subfamily 4 group A member 2 variant TINUR; nuclear receptor subfamily 4 group A member 2 variant NURR1a; nuclear receptor subfamily 4 group A member 2 variant NURR1b; nuclear receptor subfamily 4 group A member 2 variant NURR1c; nuclear receptor subfamily 4 group A member 2 variant NURR2c; nur related protein 1; NUR-related factor 1; Orphan nuclear receptor NURR1; Regenerating liver nuclear receptor 1; RNR-1; SL-322
Gene Aliases: HZF-3; NOT; Nr4a2; Nurr1; RNR-1; Rnr1; TINOR; TINUR
UniProt ID: (Mouse) Q06219, (Rat) Q07917
Entrez Gene ID: (Mouse) 18227, (Rat) 54278
Molecular Function:
C4 zinc finger nuclear receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support