Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Ocular albinism type 1 protein is a conserved integral membrane protein with seven transmembrane domains. It is expressed in the eye and epidermal melanocytes. Defects in GPR143 are the cause of ocular albinism type 1 (OA1); also known as Nettleship-Falls type ocular albinism. OA1 is an X-linked disorder characterized by severe impairment of visual acuity, retinal hypopigmentation and the presence of macromelanosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-protein coupled receptor 143; homolog of human ocular albinism 1 (Nettleship-Falls); MOA1; ocular albinism 1; Ocular albinism type 1 protein; Ocular albinism type 1 protein homolog
Gene Aliases: GPR143; NYS6; OA1; RGD1565799
UniProt ID: (Human) P51810, (Mouse) P70259
Entrez Gene ID: (Human) 4935, (Mouse) 18241, (Rat) 302619
Molecular Function:
G-protein coupled receptor
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support