Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MMDLRNTPAK SLDKFIEDYL LPDTCFRMQI NHAIDIICGF LKERCFRGSS YPVCVSKVVK GGSSGKGTTL RGRSDADLVV FLSPLTTFQD QLNRRGEFIQ EIRRQLEACQ RERAFSVKFE VQAPRWGNPR ALSFVLSSLQ LGEGVEFDVL PAFDALGQLT GSYKPNPQIY VKLIEECTDL QKEGEFSTCF TELQRDFLKQ RPTKLKSLIR LVKHWYQNCK KKLGKLPPQY ALELLTVYAW ERGSMKTHFN TAQGFRTVLE LVINYQQLCI YWTKYYDFKN PIIEKYLRRQ LTKPRPVILD PADPTGNLGG GDPKGWRQLA QEAEAWLNYP CFKNWDGSPV SSWILLVRPP ASSLPFIPAP LHEA
OAS1 is an interferon inducible protein that may play a role in mediating resistance to virus infection, control of cell growth, differentiation, and apoptosis. It binds double-stranded RNA and polymerizes ATP into PPP(A2'P5'A)N oligomers, which activate the latent RNase L that, when activated, cleaves single-stranded RNAs. This protein is associated with different subcellular fractions such as mitochondrial, nuclear, and rough/smooth microsomal fractions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (2-5')oligo(A) synthase 1; 2',5'-oligo A synthetase 1; 2'-5'-oligoadenylate synthase 1; 2'-5'-oligoadenylate synthetase 1, 40/46kDa; 2'-5'-oligoisoadenylate synthetase 1; 2-5A synthase 1; 2-5A synthetase 1; E16 (2'-5') oligo A synthetase; E18/E16; IFI 4; IFI4; OAS 1; p46/p42 OAS
Gene Aliases: E18/E16; IFI-4; OAS1; OIAS; OIASI
UniProt ID: (Human) P00973
Entrez Gene ID: (Human) 4938
Molecular Function: nucleotidyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support