Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 5
Immunogen sequence: AASCYDHPCF LRGMGDPVQS WKGPGLPRAG CSGLGHPIQL DPNQKTPENS KSLNAVYPRA GSKPPSCPAP GPTGAASIVP SVPGMALDLS QIPTKELDRF IQDHLKPSPQ FQEQVKKAID IILRCLHENC VHKASRVSKG GSFGRGTDLR DGCDVELIIF LNCFTDYKDQ GPRRAEILDE MRAQLESWWQ DQVPSLSLQF PEQNVPEALQ FQLVSTALKS WTDVSLLPAF DAVGQLSSGT KPNPQVYSRL LTSGCQEGEH KACFAELRRN FMNIRPVKLK NLILLVKHWY RQVAAQNKGK G; Positive Samples: U-87MG, DU145, HeLa; Cellular Location: Cytoplasm, Nucleus
OAS3 is an interferon inducible protein that belongs to the 2-5A synthetase family, may play a role in mediating resistance to virus infection, control of cell growth, differentiation, and apoptosis. OAS3 synthesizes preferentially dimeric 2',5'-oligoadenylate molecules. GTP can be an alternative substrate. OAS3 binds double-stranded RNA and polymerizes ATP into PPP(A2'P5'A)N oligomers, which activate the latent RNase L that, when activated, cleaves single-stranded RNAs. The protein is present at high level in placenta trophoblast.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (2-5')oligo(A) synthase 3; (2-5')oligo(A) synthetase 3; 2',5'-oligoadenylate synthetase-like 10; 2'-5' oligoadenylate synthetase-like 10; 2'-5'-oligoadenylate synthase 3; 2'-5'-oligoadenylate synthetase 3, 100kDa; 2'-5'oligoadenylate synthetase p100; 2-5A synthase 3; 2-5A synthetase 3; p100 OAS; p100OAS
Gene Aliases: OAS3; Oasl10; P/OKcl.4; p100; p100OAS
UniProt ID: (Human) Q9Y6K5, (Rat) Q5MYT7, (Mouse) Q8VI93
Entrez Gene ID: (Human) 4940, (Rat) 494202, (Mouse) 246727
Molecular Function:
nucleotidyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support