Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total SLCO1A2.
The organic anion transporting polypeptides, OATP-A (also designated OATP1, OATP1A2 and SLC21A3) and OATP-C (also designated OATP2, SLC21A6 and LST1), mediate hepatic uptake of cardiac glycosides. The expression of OATP-C, but not OATP-A, is inducible by phenobarbital and pregnenolone-16a-carbonitrile, resulting in the increased capacity of the liver to extract cardiac glycosides from the plasma. OATP-A is expressed in liver and kidney and helps mediate sodium-independent uptake of the anionic steroid conjugates dehydroepiandrosterone sulfate, estradiol-17-glucuronide and prostaglandin. OATP-C is exclusively expressed in liver and is localized to the basolateral hepatocyte membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OATP-1; OATP-A; OATP1A2; organic anion transporting polypeptide A; Organic anion-transporting polypeptide 1; SO1A2; Sodium-independent organic anion transporter; solute carrier family 21 (organic anion transporter), member 3; Solute carrier family 21 member 3; Solute carrier organic anion transporter family member 1A2; solute carrier organic anion transporter family, member 1A2
Gene Aliases: OATP; OATP-A; OATP1; OATP1A2; SLC21A3; SLCO1A2
UniProt ID: (Human) P46721
Entrez Gene ID: (Human) 6579
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support