Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Major protein of the otoconia, a calcium carbonate structure in the saccule and utricle of the ear. Together with OTOL1, acts as a scaffold for otoconia biomineralization: sequesters calcium and forms interconnecting fibrils between otoconia that are incorporated into the calcium crystal structure. Together with OTOL1, modulates calcite crystal morphology and growth kinetics. It is unlikely that this protein has phospholipase A2 activity. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Oc90; Otoconin-90; Phospholipase A2 homolog
Gene Aliases: OC90; PLA2L
UniProt ID: (Human) Q02509
Entrez Gene ID: (Human) 729330
Molecular Function:
phospholipase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support