Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This gene encodes the human homologue of the mouse p (pink-eyed dilution) gene. The encoded protein is believed to be an integral membrane protein involved in small molecule transport, specifically tyrosine - a precursor of melanin. Mutations in this gene result in type 2 oculocutaneous albinism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eye color 2 (central brown); eye color 3 (brown); hair color 3 (brown); Melanocyte-specific transporter protein; oculocutaneous albinism II (pink-eye dilution homolog, mouse); P protein; P-protein; Pink-eyed dilution protein homolog; total brown iris pigmentation
Gene Aliases: BEY; BEY1; BEY2; BOCA; D15S12; EYCL; EYCL2; EYCL3; HCL3; OCA2; P; PED; SHEP1
UniProt ID: (Human) Q04671
Entrez Gene ID: (Human) 4948
Molecular Function:
primary active transporter
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support