Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects OCIAD1 with an apparent molecular weight of ~35 kDa as has been demonstrated by several researches.
Immunogen sequence: MNGRADFRE PNAEVPRPIP HIGPDYIPTE EERRVFAECN DESFWFRSVP LAATSMLITQ GLISKGILSS HPKYGSIPKL ILACIMGYFA GKLSYVKTCQ EKFKKLENSP LGEALRSGQA RRSSPPGHYY QKSKYDSSVS GQSSFVTSPA ADNIEMLPHY EPIPFSSSMN ESAPTGITDH IVQGPDPNLE ESPKRKNITY EELRNKNRES YEVSLTQKTD PSVRPMHERV PKKEVKVNKY GDTWDE (1-245 aa encoded by B C003409)
Maintains stem cell potency. Increases STAT3 phosphorylation and controls ERK phosphorylation. May act as a scaffold, increasing STAT3 recruitment onto endosomes. Involved in integrin-mediated cancer cell adhesion and colony formation in ovarian cancer (PubMed:20515946). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: expressed during mesenchymal induction 2; FLJ20455; induced during mesenchymal induction 2; MGC111072; OCIA domain-containing protein 1; Ovarian cancer immunoreactive antigen domain containing 1; Ovarian carcinoma immunoreactive antigen
Gene Aliases: 6030432N09Rik; AI481327; ASRIJ; AW557942; B230209J16Rik; BB021357; Emi2; Imi2; OCIA; OCIAD1; TPA018
UniProt ID: (Human) Q9NX40, (Rat) Q5XIG4, (Mouse) Q9CRD0
Entrez Gene ID: (Human) 54940, (Rat) 289590, (Mouse) 68095
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support