Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
OCT1 (POU2F1) is a ubiquitously expressed transcription factor that binds to the octamer motif (5'-ATTTGCAT-3') and activates the promoters of the genes for some small nuclear RNAs (snRNA) and of genes such as those for histone H2B and immunoglobulins. It modulates transcription transactivation by NR3C1, AR and PGR. In case of human herpes simplex virus (HSV) infection, POU2F1 forms a multiprotein-DNA complex with the viral transactivator protein VP16 and HCFC1 thereby enabling the transcription of the viral immediate early genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NF-A1; Oct-1; Octamer-binding protein 1; Octamer-binding transcription factor 1; Octamer-binding transcription factor-1; OTF-1; POU domain, class 2, transcription factor 1; ubiquitous transcription factor
Gene Aliases: 2810482H01Rik; NF-A1; Oct-1; oct-1B; OCT1; Otf-1; OTF1; POU2F1
UniProt ID: (Human) P14859, (Rat) P31503, (Mouse) P25425
Entrez Gene ID: (Human) 5451, (Rat) 171068, (Mouse) 18986
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support