Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPYVFSFKMP QEQGQMFQYY PVYMVLPWEQ PQQTVPRSPQ QTRQQQYEEQ IPFYAQFGYI PQLAEPAISG GQQQLAFDPQ LGTAPEIAVM STGEEIPYLQ KEAINFRHDS AGVFMPSTSP KPSTTNVFTS AVDQTITPEL PEEKDKTDSL REP
Tooth-associated epithelia protein that probably plays a role in odontogenesis, the complex process that results in the initiation and generation of the tooth. May be incorporated in the enamel matrix at the end of mineralization process. Involved in the induction of RHOA activity via interaction with ARHGEF and expression of downstream factors such as ROCK. Plays a role in attachment of the junctional epithelium to the tooth surface.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apin; Odontogenic ameloblast-associated protein
Gene Aliases: APIN; ODAM
UniProt ID: (Human) A1E959
Entrez Gene ID: (Human) 54959
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support