Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SLETKIAKW NLQSRMNKNE AIVMKEASRQ KTVALKKGI (78-269 aa encoded by BC009779)
ODF2L, also named as KIAA1229, has many isoforms with MW 60-80 kDa. The function of ODF2L (Outer dense fiber protein 2-like) remains largely unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basal body centriole-associated protein; ODF2-like protein; outer dense fiber of sperm tails 2-like; Outer dense fiber protein 2-like; Protein BCAP
Gene Aliases: 4733401D09Rik; 9630045K08Rik; BCAP; D3Ertd250e; dJ977L11.1; KIAA1229; ODF2L; RGD1308397
UniProt ID: (Human) Q9ULJ1, (Mouse) Q9D478
Entrez Gene ID: (Human) 57489, (Mouse) 52184, (Rat) 685425
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support