Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LYSSPGPKY LIPPTTGFMK HTPTKLRAPA YSFRGAPMLL AENCSPGPRY NVNPKILRTG KDLGPAYSIL GRYQTKTMLT PG (22-102 aa encoded by BC126248)
Outer dense fibers are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail. May help to maintain the passive elastic structures and elastic recoil of the sperm tail.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 135; Ciliary microtubule associated protein 1A; Outer dense fiber of sperm tails protein 3; Outer dense fiber protein 3; Sperm tail protein SHIPPO 1; sperm tail protein SHIPPO1; Transcript induced in spermiogenesis protein 50
Gene Aliases: CIMAP1A; CT135; ODF3; SHIPPO1; TISP50
UniProt ID: (Human) Q96PU9
Entrez Gene ID: (Human) 113746, (Rat) 365387
Molecular Function:
structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support