Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MAAQPLRHRS RCATPPRGDF CGGTERAIDQ ASFTTSMEWD TQVVKGSSPL GPAGLGAEEP AAGPQLPSWL QPERCAVFQC AQCHAVLADS VHLAWDLSRS LGAVVFSRVT NNVVLEAPFL VGIEGSLKGS TYNLLFCGSC GIPVGFHLYS THAALAALRG HFCLSSDKMV CYLLKTKAIV NASEMDIQNV PLSEKIAELK EKIVLTHNRL KSLMKILSEV TPDQSKPEN
Required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 86; CT86; LAP2alpha interactor-25; MIS18 kinetochore protein homolog B; OIP-5; Opa-interacting protein 5; Protein Mis18-beta
Gene Aliases: 5730547N13Rik; CT86; hMIS18beta; LINT-25; MIS18B; MIS18beta; OIP5
UniProt ID: (Human) O43482
Entrez Gene ID: (Human) 11339
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support