Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May be involved in hepatocellular function and development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Liver-specific zona pellucida domain-containing protein; liver-specific ZP domain-containing protein; Oncoprotein-induced transcript 3 protein
Gene Aliases: EF-9; LZP; OIT3; UNQ826/PRO1753
UniProt ID: (Human) Q8WWZ8, (Mouse) Q8R4V5, (Rat) Q6V0K7
Entrez Gene ID: (Human) 170392, (Mouse) 18302, (Rat) 294559
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support