Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in rat.
Specificity of this antibody: OLIG1.
OLIG1 (oligodendrocyte transcription factor 1) promotes formation and maturation of oligodendrocytes - especially in the brain. It cooperates with OLIG2 to establish the pMN domain of the embryonic neural tube.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Olg-1 bHLH protein; olig2; olig3; Oligo1; Oligo2 antibody; Oligodendrocyte transcription factor 1; Oligodendrocyte-specific bHLH transcription factor 1
Gene Aliases: AI836478; AW494459; Bhlhb6; bHLHe21; Olg-1; Olg1; Olig1; Oligo1
UniProt ID: (Mouse) Q9JKN5, (Rat) Q9WUQ3
Entrez Gene ID: (Mouse) 50914, (Rat) 60394
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support