Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
OLIG1 (oligodendrocyte transcription factor 1) promotes formation and maturation of oligodendrocytes - especially in the brain. It cooperates with OLIG2 to establish the pMN domain of the embryonic neural tube.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basic domain, helix-loop-helix protein, class B, 6; bHLHb6; bHLHe21; Class B basic helix-loop-helix protein 6; Class E basic helix-loop-helix protein 21; Olg-1 bHLH protein; olig2; olig3; Oligo1; Oligo2 antibody; oligodendrocyte lineage transcription factor 1; Oligodendrocyte transcription factor 1; Oligodendrocyte-specific bHLH transcription factor 1
Gene Aliases: AI836478; AW494459; BHLHB6; BHLHE21; Olg-1; Olg1; OLIG1; Oligo1
UniProt ID: (Human) Q8TAK6, (Mouse) Q9JKN5, (Rat) Q9WUQ3
Entrez Gene ID: (Human) 116448, (Mouse) 50914, (Rat) 60394
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support