Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDSDASLVS SRPSSPEPDD LFLPARSKGS SGSAFTGGTV SSSTPSDCPP ELSAELRGAM GSAGAHPGDK LGGSGFKSSS SSTSSSTSSA AASSTKKDKK QMTEPELQQL RLKINSRERK RMHDLNIAMD GLREVMPYAH GPSVRKLSKI TTLLLARNYI LMLTNSLEEM KRLVSEIYGG HHAGFHPSAC GGLAHSAPLP AATAHPAAAA HAAHHPAVHH PILPPAAAAA AAAAAAAAVS SASLPGSGLP SVGSIRPPHG LLKSPSAAAA APLGGGGGGS GASGGFQHWG GMPCPCSMCQ VPPPHHHVSA MGAGSLPRLT SDAK (1-323 aa encoded by BC047511)
This gene encodes a basic helix-loop-helix transcription factor which is expressed in oligodendroglial tumors of the brain. The protein is an essential regulator of ventral neuroectodermal progenitor cell fate. The gene is involved in a chromosomal translocation t(14;21)(q11.2;q22) associated with T-cell acute lymphoblastic leukemia. Its chromosomal location is within a region of chromosome 21 which has been suggested to play a role in learning deficits associated with Down syndrome. Tissue specificity: Expressed in the brain, in oligodendrocytes. Strongly expressed in oligodendrogliomas, while expression is weak to moderate in astrocytomas. Expression in glioblastomas highly variable.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basic domain, helix-loop-helix protein, class B, 1; Basic helix loop helix protein class B1 (bHLHB1); basic helix-loop-helix protein 19 (bHLHe19); bHLHb1; bHLHe19; Class B basic helix-loop-helix protein 1; Class E basic helix-loop-helix protein 19; human protein kinase C-binding protein RACK17; olig-2; Oligo2; Oligo2 antibody; Oligodendrocyte specific bHLH transcription factor 2; Oligodendrocyte transcription factor 2; oligodendrocyte-specific bHLH transcription factor 2; Protein kinase C-binding protein 2; Protein kinase C-binding protein 2 (PRKCBP2); Protein kinase C-binding protein RACK17
Gene Aliases: AI604895; BHLHB1; BHLHE19; Olg-2; OLIG2; OLIGO2; PRKCBP2; RACK17; RK17
UniProt ID: (Human) Q13516, (Mouse) Q9EQW6
Entrez Gene ID: (Human) 10215, (Mouse) 50913, (Rat) 304103
Molecular Function: basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support