Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western blots, no cross-reactivity with recombinant human (rh) Olig1 or rhOlig2 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
OLIG3 may determine the distinct specification program of class A neurons in the dorsal part of the spinal cord and suppress specification of class B neurons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basic helix-loop-helix domain-containing class B protein 7; bHLHb7; bHLHe20; bHLHe20oligo3; Class B basic helix-loop-helix protein 7; Class E basic helix-loop-helix protein 20; Oligo3; Oligodendrocyte transcription factor 3; Oligodendrocyte-specific bHLH transcription factor 3
Gene Aliases: BHLHB7; BHLHE20; OLIG3
UniProt ID: (Human) Q7RTU3, (Mouse) Q6PFG8
Entrez Gene ID: (Human) 167826, (Mouse) 94222
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support