Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTFDDLKIQT VKDQPDEKSN GKKAKGLQFL YSPWWCLAAA TLGVLCLGLV VTIMVLGMQL SQVSDLLTQE QANLTHQKKK LEGQISARQQ AEEASQESEN ELKEMIETLA RKLNEKSKEQ MELHHQNLNL QETLKRVANC SAPCPQDWIW HGENCYLFSS GSFNWEKSQE KCLSLDAKLL KINSTADLDF IQQAISYSSF PFWMGLSRRN PSYPWLWEDG SPLMPHLFRV RGAVSQTYPS GTCAYIQRGA VYAENCILAA FSICQKKANL RAQ
LOX-1 (lectin-like oxidized LDL receptor -1) is a 40-50 kDa type II membrane glycoprotein class E scavenger receptor expressed by endothelial cells, macrophages, activated platelets and smooth muscle cells. LOX-1 binds to a wide range of ligands, including oxidized low-density lipoprotein (oxLDL), hypochlorite modified high-density lipoprotein (HDL), aged /apoptotic cells, activated platelets and bacteria, reflecting its versatile physiological functions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type lectin domain family 8 member A; hLOX 1; hLOX-1; hLOX1; Lectin-like oxidized LDL receptor 1; Lectin-type oxidized LDL receptor 1; LOX 1; LOX-1; ox LDL receptor 1; Ox-LDL receptor 1; oxidized low density lipoprotein (lectin-like) receptor 1; Oxidized low-density lipoprotein receptor 1; oxidized low-density lipoprotein receptor 1, soluble form; scavenger receptor class E, member 1; sLOX 1; soluble LOX 1
Gene Aliases: CLEC8A; LOX1; LOXIN; OLR1; SCARE1; SLOX1
UniProt ID: (Human) P78380
Entrez Gene ID: (Human) 4973
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support