Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OPA1 is a dynamin-related GTPase that is critical for the maintenance of mitochondrial morphology and mtDNA. The most commonly associated phenotype with OPA1 mutations is heterozygous optic atrophy, a heterozygous dominant trait that causes reduced visual clarity and sometimes blindness. The disease usually begins in childhood and increases in severity throughout the life of affected individuals. Usually, this phenotype is attributed to the degeneration of optic nerve fibers. Interestingly, the same type of nerve degeneration seems to be partially causative of certain schizophrenia characteristics. OPA1 dysfunction also seems to be implicated in this case; mitochondrial networks associated with critical nerves seem to link schizophrenia and OPA1. The dysfunction is associated with issues with apoptosis and normal cellular metabolic regulation, all regulated through OPA1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dynamin-like 120 kDa protein, mitochondrial; Dynamin-like GTPase OPA1, mitochondrial; dynamin-like guanosine triphosphatase; Large GTP-binding protein; LargeG; mitochondrial dynamin-like GTPase; mitochondrial OPA1; opa1 {ECO:0000312|EMBL:AAI11072.1}; optic atrophy 1 (autosomal dominant); optic atrophy 1 homolog; optic atrophy 1-like protein; Optic atrophy protein 1; Optic atrophy protein 1 homolog; RN protein
Gene Aliases: 1200011N24Rik; AI225888; AI847218; BERHS; KIAA0567; largeG; lilr3; MGM1; mKIAA0567; MTDPS14; NPG; NTG; OPA1; RCJMB04_1m16
UniProt ID: (Chicken) Q5F499, (Human) O60313, (Rat) Q2TA68, (Mouse) P58281
Entrez Gene ID: (Chicken) 424900, (Rabbit) 100344494, (Human) 4976, (Rat) 171116, (Mouse) 74143
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support