Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TGVPVRSGDA TFPKAMDNVT VRQGESATLR CTIDDRVTRV AWLNRSTILY AGNDKWSIDP RVIILVNTPT QYSIMIQNVD VYDEGPYTCS VQTDNHPKTS RVHLIVQVPP QIMNISSDIT VNEGSSVTLL CLAIGRPEPT VTWRHLSVKE GQGFVSEDEY LEISDIKRDQ SGEYECSALN DVAAPDVRKV KITVNYPPYI SKAKNTGVSV GQKGILSCEA SAVPMAEFQW FKEETRLATG LDGMRIENKG RMSTLTFFNV SEKDYGNYTC VATNKLGNTN ASITLYGPGA VIDGVNSASR ALACLWLSGT LLAHFFIKF
Binds opioids in the presence of acidic lipids; probably involved in cell contact.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IgLON family member 1; OBCAM; opioid binding cell adhesion molecule; opioid binding protein/cell adhesion molecule-like preprotein; opioid-binding cell adhesion molecule; Opioid-binding protein/cell adhesion molecule; opioid-binding protein/cell adhesion molecule-like
Gene Aliases: 2900075O15Rik; 3732419F12; AI844366; B930023M13Rik; C230027C17; Gm181; IGLON1; OBCAM; OPCM; OPCML
UniProt ID: (Human) Q14982, (Rat) P32736
Entrez Gene ID: (Human) 4978, (Rat) 116597, (Mouse) 330908
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support