Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Visual pigments are the light-absorbing molecules that mediate vision. They consist of an apoprotein, opsin, covalently linked to cis-retinal.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: opsin 1 (cone pigments), short-wave-sensitive, 2; Opsin SWS-1; Opsin-1, short-wave-sensitive 1; seven transmembrane light receptor; short-wavelength-sensitive-1 cone opsin; Ultraviolet cone photoreceptor pigment; Ultraviolet-sensitive opsin
Gene Aliases: opn1sw1; opn1sw2; SWS1; uvops; zfuv
UniProt ID: (Zebrafish) Q9W6A9
Entrez Gene ID: (Zebrafish) 30582
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support