Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: Specific human olfactory receptor Olfactory receptor OR11H4.
Putative odorant receptor. Multi-pass membrane protein. Belongs to the G-protein coupled receptor 1 family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Olfactory receptor 11H4; Olfactory receptor OR14-36; olfactory receptor, family 11, subfamily H, member 4
Gene Aliases: OR11H4; OR14-36
UniProt ID: (Human) Q8NGC9
Entrez Gene ID: (Human) 390442
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support