Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FLCQTGKQAK ISMGEENQTF VSKFIFLGLS QDLQ
Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 50%.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Olfactory receptor 2D3; Olfactory receptor OR11-89; olfactory receptor, family 2, subfamily D, member 3
Gene Aliases: OR11-89; OR2D3
UniProt ID: (Human) Q8NGH3
Entrez Gene ID: (Human) 120775
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support