Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Poitive Samples: Human plasma
Immunogen sequence: MALSWVLTVL SLLPLLEAQI PLCANLVPVP ITNATLDRIT GKWFYIASAF RNEEYNKSVQ EIQATFFYFT PNKTEDTIFL REYQTRQDQC IYNTTYLNVQ MALSWVLTV LSLLPLLEAQ IPLCANLVPV PITNATLDRI TGKWFYIASA FRNEEYNKSV QEIQATFFYF TPNKTEDTIF LREYQTRQNQ CFYNSSYLNV Q
ORM1 is an acute phase plasma protein synthesized by the liver. The protein is believed to regulate the interaction between blood cells and endothelial cells, and together with haptoglobin and C reactive protein, also regulates the extravasation of the cells during infection and inflammation. ORM1 is a major acute phase reactant, both in human and in rat. In human plasma, ORM1 (44 kDa) is found at levels of 0.5-1.4 mg/mL. Its concentration in blood plasma is elevated during acute inflammation, such as trauma, malignancies, myocardial infarction, rheumatoid arthritis, bacterial infections, after major surgery, in cases of chronic pain etc, and can be used for the diagnosis of inflammatory conditions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AGP 1; Alpha-1-acid glycoprotein 1; epididymis secretory sperm binding protein Li 153w; OMD 1; Orosomucoid-1
Gene Aliases: AGP-A; AGP1; HEL-S-153w; ORM; ORM1
UniProt ID: (Human) P02763
Entrez Gene ID: (Human) 5004
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support