Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNGEEEFFDA VTGFDSDNSS GEFSEANQKV TGMIDLDTSK NNRIGKTGER PSQENGIQKH RT
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. This encoded protein contains only the sterol-binding domain. In vitro studies have shown that the encoded protein can bind strongly to phosphatic acid and weakly to phosphatidylinositol 3-phosphate, but cannot bind to 25-hydroxycholesterol. The protein associates with the Golgi apparatus. Transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20223; MGC4307; MGC8342; ORP-2; OSBP-related protein 2; oxysterol binding protein-like 2; Oxysterol-binding protein-related protein 2; RP11-157P1.2
Gene Aliases: DFNA67; DNFA67; KIAA0772; ORP-2; ORP2; OSBPL2
UniProt ID: (Human) Q9H1P3
Entrez Gene ID: (Human) 9885
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support